Product Details

SNP ID
rs139045137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75539300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCTCCCTGCTGCGGGTTAGT[A/G]CCCATCCTTGGAGTCAGCTTACCTT
Phenotype
MIM: 601767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIP1 PubMed Links

Gene Details

Gene
HIP1
Gene Name
huntingtin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243198.2 Intron NP_001230127.1
NM_005338.6 Intron NP_005329.3
XM_005250304.2 Intron XP_005250361.1
XM_005250305.2 Intron XP_005250362.1
XM_011516116.2 Intron XP_011514418.1
XM_017012099.1 Intron XP_016867588.1

View Full Product Details