Product Details

SNP ID
rs76760929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75996650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCACGATTGGTCCTGGGCCCTT[G/T]CCACTTGGATCAGAGGCAGCTTTCA
Phenotype
MIM: 124015 MIM: 616695 MIM: 616550
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POR PubMed Links

Gene Details

Gene
POR
Gene Name
cytochrome p450 oxidoreductase
There are no transcripts associated with this gene.

Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317785.1 Intron NP_001304714.1
NM_001317786.1 Intron NP_001304715.1
NM_001317787.1 Intron NP_001304716.1
NM_001317788.1 Intron NP_001304717.1
NM_001317789.1 Intron NP_001304718.1
NM_016086.2 Intron NP_057170.1
XM_011516292.2 Intron XP_011514594.1
XM_011516293.2 Intron XP_011514595.1
XM_011516300.1 Intron XP_011514602.1
XM_017012296.1 Intron XP_016867785.1
XM_017012297.1 Intron XP_016867786.1
XM_017012298.1 Intron XP_016867787.1
XM_017012299.1 Intron XP_016867788.1
XM_017012300.1 Intron XP_016867789.1
XM_017012301.1 Intron XP_016867790.1
XM_017012302.1 Intron XP_016867791.1
Gene
TMEM120A
Gene Name
transmembrane protein 120A
There are no transcripts associated with this gene.

View Full Product Details