Product Details

SNP ID
rs61736643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:15075109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGCCACACTCAGTAAAGAGGA[C/T]GCGCGCCTCCTGCTGGCTGCACTGG
Phenotype
MIM: 114160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALCB PubMed Links

Gene Details

Gene
CALCB
Gene Name
calcitonin related polypeptide beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000728.3 208 Silent Mutation GAC,GAT D45D NP_000719.1

View Full Product Details