Product Details

SNP ID
rs118070560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55432255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGCATCTGCACCCAGCTAATA[C/T]AGCATTTTTCTAGACTGAAACTTGT
Phenotype
MIM: 608706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYX1C1 PubMed Links

Gene Details

Gene
DYX1C1
Gene Name
dyslexia susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033559.2 Intron NP_001028731.1
NM_001033560.1 Intron NP_001028732.1
NM_130810.3 Intron NP_570722.2
Gene
DYX1C1-CCPG1
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details