Product Details

SNP ID
rs61742972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76001888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTTGCAGGGAGATGGAGGGGG[G/T]CCCAAGCACTGCCATCAGGGAGATC
Phenotype
MIM: 123828 MIM: 601590 MIM: 613130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK3 PubMed Links

Gene Details

Gene
CDK3
Gene Name
cyclin dependent kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258.2 207 Missense Mutation GGC,GTC G44V NP_001249.1
Gene
EVPL
Gene Name
envoplakin
There are no transcripts associated with this gene.

Gene
TEN1
Gene Name
TEN1 CST complex subunit
There are no transcripts associated with this gene.

Gene
TEN1-CDK3
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details