Product Details

SNP ID
rs61741641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76076722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCATCGGGCTCATCTGGGGGC[C/T]GTCGCTGCTCTTCTCCGGGCCCTAC
Phenotype
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC7 PubMed Links

Gene Details

Gene
EXOC7
Gene Name
exocyst complex component 7
There are no transcripts associated with this gene.

Gene
GALR2
Gene Name
galanin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003857.3 1695 Missense Mutation CCG,CTG P152L NP_003848.1
XM_011525427.2 1695 Missense Mutation CCG,CTG P95L XP_011523729.1
Gene
SRP68
Gene Name
signal recognition particle 68
There are no transcripts associated with this gene.

Gene
ZACN
Gene Name
zinc activated ion channel
There are no transcripts associated with this gene.

View Full Product Details