Product Details

SNP ID
rs9897208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40993870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGTGAGGTTGAGGGTCTCCAG[A/G]CCTGGAGTTGGCTGGCAGGAGTTGA
Phenotype
MIM: 616679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT40 PubMed Links

Gene Details

Gene
KRT40
Gene Name
keratin 40
There are no transcripts associated with this gene.

Gene
KRTAP3-2
Gene Name
keratin associated protein 3-2
There are no transcripts associated with this gene.

Gene
KRTAP3-3
Gene Name
keratin associated protein 3-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033185.2 264 Silent Mutation GGC,GGT G76G NP_149441.1

View Full Product Details