Product Details

SNP ID
rs146939021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41123589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACAGGTTGGGCGATAGCAAGT[A/G]GTGTGGCAGGAGACTCGGCCACAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-11 PubMed Links

Gene Details

Gene
KRTAP4-11
Gene Name
keratin associated protein 4-11
There are no transcripts associated with this gene.

Gene
KRTAP4-12
Gene Name
keratin associated protein 4-12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031854.2 579 Silent Mutation ACC,ACT T178T NP_114060.1

View Full Product Details