Product Details

SNP ID
rs61734608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43967080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCACAACTTCATTCCCATCAGAC[A/G]TGCCATACTGGAGAAAAGCTATACA
Phenotype
MIM: 604086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF155 PubMed Links

Gene Details

Gene
ZNF155
Gene Name
zinc finger protein 155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260486.1 1768 Intron NP_001247415.1
NM_001260487.1 1768 Intron NP_001247416.1
NM_001260488.1 1768 Intron NP_001247417.1
NM_003445.3 1768 Intron NP_003436.3
NM_198089.2 1768 Intron NP_932355.2
XM_005259215.3 1768 Intron XP_005259272.1
XM_011527278.2 1768 Intron XP_011525580.1
XM_011527279.2 1768 UTR 5 XP_011525581.1
XM_017027248.1 1768 Intron XP_016882737.1
Gene
ZNF221
Gene Name
zinc finger protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297588.1 1768 Silent Mutation ACA,ACG T526T NP_001284517.1
NM_001297589.1 1768 Silent Mutation ACA,ACG T526T NP_001284518.1
NM_013359.2 1768 Silent Mutation ACA,ACG T526T NP_037491.2
XM_017027232.1 1768 Intron XP_016882721.1
XM_017027233.1 1768 Silent Mutation ACA,ACG T468T XP_016882722.1

View Full Product Details