Product Details

SNP ID
rs45591734
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52115253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGCTGACTAAAGGAATTCCCACA[C/T]TGATTGCATTTGTAAGGTTTCTCTC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZNF616 PubMed Links

Gene Details

Gene
ZNF616
Gene Name
zinc finger protein 616
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178523.4 2194 Silent Mutation CAA,CAG Q637Q NP_848618.2
XM_017027438.1 2194 Silent Mutation CAA,CAG Q668Q XP_016882927.1

View Full Product Details