Product Details

SNP ID
rs112816154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8811229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGTGTGAGCTCTCTCAAACTAT[C/T]TTAGGGATGAAAGATCAATGAGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF558 PubMed Links

Gene Details

Gene
ZNF558
Gene Name
zinc finger protein 558
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304350.1 2376 UTR 3 NP_001291279.1
NM_144693.2 2376 UTR 3 NP_653294.1
XM_011527703.2 2376 UTR 3 XP_011526005.1
XM_017026370.1 2376 UTR 3 XP_016881859.1
XM_017026371.1 2376 UTR 3 XP_016881860.1
XM_017026372.1 2376 UTR 3 XP_016881861.1

View Full Product Details