Product Details

SNP ID
rs113891868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17436125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTAACAGAATCTCATCCATCT[A/G]GACAGCCACATTTCCCAGCCTCCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM221 PubMed Links

Gene Details

Gene
TMEM221
Gene Name
transmembrane protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190844.1 1314 UTR 3 NP_001177773.1
XM_011527603.2 1314 Intron XP_011525905.1

View Full Product Details