Product Details

SNP ID
rs61735763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29034887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGCCTTGAAGCACTATCTGAC[A/G]CCAAGATCTGAACCTCACTGTCTGG
Phenotype
MIM: 604735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP16 PubMed Links

Gene Details

Gene
USP16
Gene Name
ubiquitin specific peptidase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001992.1 494 Silent Mutation ACA,ACG T97T NP_001001992.1
NM_001032410.1 494 Silent Mutation ACA,ACG T97T NP_001027582.1
NM_006447.2 494 Silent Mutation ACA,ACG T97T NP_006438.1
XM_017028257.1 494 Silent Mutation ACA,ACG T97T XP_016883746.1
XM_017028258.1 494 Silent Mutation ACA,ACG T97T XP_016883747.1
XM_017028259.1 494 Silent Mutation ACA,ACG T97T XP_016883748.1
XM_017028260.1 494 UTR 5 XP_016883749.1
XM_017028261.1 494 UTR 5 XP_016883750.1

View Full Product Details