Product Details

SNP ID
rs61739352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39380433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCGTGGTTGCTGGTGCTCAGC[C/T]TCGTGTTTGGATGCAATGTTCTTAG
Phenotype
MIM: 602915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WRB PubMed Links

Gene Details

Gene
WRB
Gene Name
tryptophan rich basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146218.1 190 Intron NP_001139690.1
NM_004627.4 190 Missense Mutation CTC,TTC L17F NP_004618.2
Gene
WRB-SH3BGR
Gene Name
WRB-SH3BGR readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317744.1 190 Missense Mutation CTC,TTC L17F NP_001304673.1

View Full Product Details