Product Details

SNP ID
rs114677323
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35546566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATCCCAGGAGTCCGGTGCTTG[A/C]ATAAATGAAAGGATGAATGAACGGA
Phenotype
MIM: 612842
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RASD2 PubMed Links

Gene Details

Gene
RASD2
Gene Name
RASD family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014310.3 2571 Intron NP_055125.2
XM_005261442.4 2571 Intron XP_005261499.1
XM_011530040.2 2571 Intron XP_011528342.1
XM_017028701.1 2571 UTR 5 XP_016884190.1
XM_017028702.1 2571 Intron XP_016884191.1

View Full Product Details