Product Details

SNP ID
rs61745303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:95840693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCATTGCTCTGGCCTGTAAATA[C/T]AAAGGAAACGATGAGATCTGTTTGA
Phenotype
MIM: 179061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDHA2 PubMed Links

Gene Details

Gene
PDHA2
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005390.4 606 Silent Mutation TAC,TAT Y181Y NP_005381.1

View Full Product Details