Product Details

SNP ID
rs74409947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130359400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGAATTCGGCCTTCACCAGGGC[A/G]TGGATGGAAGCCTGGCCTACATTGC
Phenotype
MIM: 606587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN18 PubMed Links

Gene Details

Gene
PTPN18
Gene Name
protein tyrosine phosphatase, non-receptor type 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142370.1 379 Intron NP_001135842.1
NM_014369.3 379 Missense Mutation ATG,GTG M95V NP_055184.2
XM_006712416.3 379 Intron XP_006712479.1
XM_006712417.1 379 Missense Mutation ATG,GTG M95V XP_006712480.1
XM_017003798.1 379 Intron XP_016859287.1

View Full Product Details