Product Details

SNP ID
rs61744654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132553138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGTTCACCGTGTCTGTGTCGG[G/T]AATTTGCATCAGCGTGTCCTGGATT
Phenotype
MIM: 606927
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAAR8 PubMed Links

Gene Details

Gene
TAAR8
Gene Name
trace amine associated receptor 8
There are no transcripts associated with this gene.

View Full Product Details