Product Details

SNP ID
rs117702072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:136561380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGTATCTGTGGAGTTAGCTGA[C/T]TATCACGTGCCTACCCTGGTCTGTA
Phenotype
MIM: 602448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K5 PubMed Links

Gene Details

Gene
MAP3K5
Gene Name
mitogen-activated protein kinase kinase kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005923.3 Intron NP_005914.1
XM_011535839.2 Intron XP_011534141.2
XM_017010870.1 Intron XP_016866359.1
XM_017010871.1 Intron XP_016866360.1
XM_017010872.1 Intron XP_016866361.1
XM_017010873.1 Intron XP_016866362.1
XM_017010874.1 Intron XP_016866363.1
XM_017010875.1 Intron XP_016866364.1
XM_017010876.1 Intron XP_016866365.1
XM_017010877.1 Intron XP_016866366.1

View Full Product Details