Product Details

SNP ID
rs114430233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52691408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAGCTGCCGCCGCCTCCCGCGCC[A/G]CCGTTGACGCTCACCTGGCTGTTCT
Phenotype
MIM: 611158
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT77 PubMed Links

Gene Details

Gene
KRT77
Gene Name
keratin 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175078.2 1285 Silent Mutation GGC,GGT G498G NP_778253.2
XM_011538288.2 1285 Silent Mutation GGC,GGT G265G XP_011536590.1
XM_011538289.2 1285 Intron XP_011536591.1

View Full Product Details