Product Details

SNP ID
rs61746918
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:89185318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTTGGAAGCCCCAGGTAGAGTG[A/G]CAATCATTATGTTGCTAGCCATGTC
Phenotype
MIM: 612466 MIM: 612468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GBP4 PubMed Links

Gene Details

Gene
GBP4
Gene Name
guanylate binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052941.4 1975 Missense Mutation GCC,GTC A620V NP_443173.2
Gene
GBP7
Gene Name
guanylate binding protein 7
There are no transcripts associated with this gene.

View Full Product Details