Product Details

SNP ID
rs61747736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2799410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACATCATTGTGAACCCTGACGCA[C/G]TTGGGGTTTTACGCAATGACATTGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS41 PubMed Links

Gene Details

Gene
PRSS41
Gene Name
protease, serine 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135086.1 1039 Missense Mutation CTT,GTT L141V NP_001128558.1
XM_011522477.2 1039 Missense Mutation CTT,GTT L157V XP_011520779.1

View Full Product Details