Product Details

SNP ID
rs41270849
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:161218859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGATGGACTCCAGCTCCTGATC[A/G]CCCTTTGACTCCCATCTCCAGGGCC
Phenotype
MIM: 107670 MIM: 147139 MIM: 602985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOA2 PubMed Links

Gene Details

Gene
APOA2
Gene Name
apolipoprotein A2
There are no transcripts associated with this gene.

Gene
FCER1G
Gene Name
Fc fragment of IgE receptor Ig
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004106.1 Intron NP_004097.1
Gene
MIR5187
Gene Name
microRNA 5187
There are no transcripts associated with this gene.

Gene
NDUFS2
Gene Name
NADH:ubiquinone oxidoreductase core subunit S2
There are no transcripts associated with this gene.

Gene
TOMM40L
Gene Name
translocase of outer mitochondrial membrane 40 like
There are no transcripts associated with this gene.

View Full Product Details