Product Details

SNP ID
rs45458194
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:171339249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTCTATTTGGACTGATCAGA[C/T]GTTGTTCTCACATCAAATCCAAGAA
Phenotype
MIM: 136131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO4 PubMed Links

Gene Details

Gene
FMO4
Gene Name
flavin containing monooxygenase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002022.2 1707 Intron NP_002013.1
XM_005245045.3 1707 Intron XP_005245102.1
XM_005245046.3 1707 Intron XP_005245103.1
XM_005245048.3 1707 Intron XP_005245105.1
XM_006711243.3 1707 Intron XP_006711306.1
XM_011509347.2 1707 Missense Mutation ACG,ATG T425M XP_011507649.1
XM_011509348.2 1707 Intron XP_011507650.1
XM_011509349.2 1707 Intron XP_011507651.1
Gene
TOP1P1
Gene Name
topoisomerase (DNA) I pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details