Product Details

SNP ID
rs41265201
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:193059916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGCGAAACTATCGCTCTTCCC[A/C]GTCCCGCTTCCGCGCCTGTCCACCC
Phenotype
MIM: 600063 MIM: 610667
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCARNA18B PubMed Links

Gene Details

Gene
SCARNA18B
Gene Name
small Cajal body-specific RNA 18B
There are no transcripts associated with this gene.

Gene
TROVE2
Gene Name
TROVE domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042369.2 192 UTR 5 NP_001035828.1
NM_001042370.2 192 UTR 5 NP_001035829.2
NM_001173524.1 192 Intron NP_001166995.1
NM_001173525.1 192 UTR 5 NP_001166996.1
NM_004600.5 192 UTR 5 NP_004591.2
XM_006711495.3 192 Intron XP_006711558.1
XM_006711496.3 192 UTR 5 XP_006711559.1
XM_006711497.3 192 UTR 5 XP_006711560.1
XM_011509922.2 192 UTR 5 XP_011508224.1
XM_017002180.1 192 Intron XP_016857669.1
XM_017002181.1 192 UTR 5 XP_016857670.1
XM_017002182.1 192 Intron XP_016857671.1
XM_017002183.1 192 Intron XP_016857672.1
Gene
UCHL5
Gene Name
ubiquitin C-terminal hydrolase L5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199261.1 192 Intron NP_001186190.1
NM_001199262.1 192 Intron NP_001186191.1
NM_001199263.1 192 Intron NP_001186192.1
NM_015984.3 192 Intron NP_057068.1
XM_005245244.4 192 Intron XP_005245301.1
XM_005245246.4 192 Intron XP_005245303.1
XM_005245247.4 192 Intron XP_005245304.1
XM_005245248.4 192 Intron XP_005245305.1
XM_005245249.4 192 Intron XP_005245306.1
XM_006711366.3 192 Intron XP_006711429.1
XM_006711367.3 192 Intron XP_006711430.1
XM_006711368.3 192 Intron XP_006711431.1
XM_006711369.2 192 Intron XP_006711432.1
XM_006711370.3 192 Missense Mutation CAG,CCG Q28P XP_006711433.3
XM_006711371.3 192 Intron XP_006711434.1
XM_011509604.1 192 Intron XP_011507906.1
XM_011509607.2 192 Intron XP_011507909.1
XM_011509608.2 192 Intron XP_011507910.1
XM_011509609.2 192 Intron XP_011507911.1
XM_017001429.1 192 Intron XP_016856918.1
XM_017001430.1 192 Intron XP_016856919.1
XM_017001431.1 192 Intron XP_016856920.1
XM_017001432.1 192 Intron XP_016856921.1
XM_017001433.1 192 Intron XP_016856922.1
XM_017001434.1 192 Missense Mutation CAG,CCG Q28P XP_016856923.1
XM_017001435.1 192 Missense Mutation CAG,CCG Q28P XP_016856924.1
XM_017001436.1 192 Intron XP_016856925.1
XM_017001437.1 192 Intron XP_016856926.1
XM_017001438.1 192 Intron XP_016856927.1
XM_017001439.1 192 UTR 5 XP_016856928.1
XM_017001440.1 192 UTR 5 XP_016856929.1
XM_017001441.1 192 UTR 5 XP_016856930.1
XM_017001442.1 192 Intron XP_016856931.1
XM_017001443.1 192 Intron XP_016856932.1
XM_017001444.1 192 UTR 5 XP_016856933.1

View Full Product Details