Product Details

SNP ID
rs45500702
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:197508630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCCTCTTCTAGGATTTTAGCT[A/G]AAATTATATTCTTTCAATAAATATT
Phenotype
MIM: 613292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND1B PubMed Links

Gene Details

Gene
DENND1B
Gene Name
DENN domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195215.1 3970 UTR 3 NP_001182144.1
NM_001195216.1 3970 Intron NP_001182145.1
NM_001300858.1 3970 Intron NP_001287787.1
NM_144977.4 3970 Intron NP_659414.2
XM_005244931.2 3970 UTR 3 XP_005244988.1
XM_006711192.2 3970 UTR 3 XP_006711255.1
XM_006711193.3 3970 UTR 3 XP_006711256.1
XM_006711194.3 3970 UTR 3 XP_006711257.1
XM_011509246.2 3970 UTR 3 XP_011507548.1
XM_011509247.1 3970 UTR 3 XP_011507549.1
XM_011509248.2 3970 UTR 3 XP_011507550.1
XM_011509249.2 3970 UTR 3 XP_011507551.1
XM_011509251.2 3970 Intron XP_011507553.1
XM_017000470.1 3970 UTR 3 XP_016855959.1
XM_017000471.1 3970 UTR 3 XP_016855960.1
XM_017000472.1 3970 UTR 3 XP_016855961.1

View Full Product Details