Product Details

SNP ID
rs41264995
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:155947598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCCAGTAGTGTTCAAGGACAAG[C/G]CCCTCTGATCCCTATGGCTTGGTTC
Phenotype
MIM: 607560 MIM: 609043
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF2 PubMed Links

Gene Details

Gene
ARHGEF2
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162383.1 3497 UTR 3 NP_001155855.1
NM_001162384.1 3497 UTR 3 NP_001155856.1
NM_004723.3 3497 UTR 3 NP_004714.2
XM_005245587.2 3497 UTR 3 XP_005245644.1
XM_005245588.2 3497 UTR 3 XP_005245645.1
XM_005245589.4 3497 UTR 3 XP_005245646.1
XM_005245590.3 3497 UTR 3 XP_005245647.1
XM_005245591.3 3497 UTR 3 XP_005245648.1
XM_005245592.2 3497 UTR 3 XP_005245649.1
XM_005245593.4 3497 UTR 3 XP_005245650.1
XM_005245594.2 3497 UTR 3 XP_005245651.1
XM_006711622.3 3497 UTR 3 XP_006711685.1
XM_006711623.3 3497 UTR 3 XP_006711686.1
XM_006711624.3 3497 UTR 3 XP_006711687.1
XM_006711625.3 3497 UTR 3 XP_006711688.1
XM_006711626.2 3497 UTR 3 XP_006711689.1
XM_011510136.2 3497 UTR 3 XP_011508438.2
XM_011510137.2 3497 UTR 3 XP_011508439.2
XM_011510139.2 3497 UTR 3 XP_011508441.1
XM_011510141.2 3497 UTR 3 XP_011508443.1
XM_011510143.2 3497 UTR 3 XP_011508445.1
XM_017002794.1 3497 UTR 3 XP_016858283.1
XM_017002795.1 3497 UTR 3 XP_016858284.1
XM_017002796.1 3497 UTR 3 XP_016858285.1
XM_017002797.1 3497 UTR 3 XP_016858286.1
XM_017002798.1 3497 UTR 3 XP_016858287.1
XM_017002799.1 3497 UTR 3 XP_016858288.1
Gene
MIR6738
Gene Name
microRNA 6738
There are no transcripts associated with this gene.

Gene
RXFP4
Gene Name
relaxin/insulin like family peptide receptor 4
There are no transcripts associated with this gene.

View Full Product Details