Product Details
- SNP ID
-
rs41265041
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:156344649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACCCTCCCCCAGCTATATTAATC[G/T]TCAAGCAAGTTCCCCACCAGCCACT
- Phenotype
-
MIM: 600114
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCT3
PubMed Links
Gene Details
- Gene
- CCT3
- Gene Name
- chaperonin containing TCP1 subunit 3
There are no transcripts associated with this gene.
- Gene
- TSACC
- Gene Name
- TSSK6 activating cochaperone
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304817.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291746.1 |
NM_001304818.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291747.1 |
NM_001304819.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291748.1 |
NM_001304820.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291749.1 |
NM_001304821.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291750.1 |
NM_001304822.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291751.1 |
NM_001304823.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291752.1 |
NM_001304824.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291753.1 |
NM_001304825.1 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_001291754.1 |
NM_001304826.1 |
293 |
Missense Mutation |
CGT,CTT |
R53L |
NP_001291755.1 |
NM_144627.4 |
293 |
Missense Mutation |
CGT,CTT |
R35L |
NP_653228.1 |
View Full Product Details