Product Details

SNP ID
rs41273539
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:158754857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAATCAACATTCCAATTATAG[A/G]AGTCTCTTGTTTCCTTTTCTGTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6K6 PubMed Links

Gene Details

Gene
OR6K6
Gene Name
olfactory receptor family 6 subfamily K member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005184.1 42 Silent Mutation AGA,AGG R14R NP_001005184.1
Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 42 Intron NP_001005185.1
XM_017000325.1 42 Intron XP_016855814.1
XM_017000326.1 42 Intron XP_016855815.1
XM_017000327.1 42 Intron XP_016855816.1

View Full Product Details