Product Details

SNP ID
rs41273541
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:158776888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGATGATGCTCCCAAAGAAGA[C/T]GAGGACCACAGCAAGATGTGAGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6N1 PubMed Links

Gene Details

Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 748 Intron NP_001005185.1
XM_017000325.1 748 Intron XP_016855814.1
XM_017000326.1 748 Intron XP_016855815.1
XM_017000327.1 748 Intron XP_016855816.1
Gene
OR6N2
Gene Name
olfactory receptor family 6 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005278.1 748 Missense Mutation NP_001005278.1

View Full Product Details