Product Details

SNP ID
rs41315595
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:227815807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTGGTGGCCCCTCCCCGGGTC[A/G]CAGCATTGGTCCACAGACAGCCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS38 PubMed Links

Gene Details

Gene
PRSS38
Gene Name
protease, serine 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183062.2 91 Missense Mutation ACA,GCA T31A NP_898885.1
XM_011544175.2 91 Missense Mutation ACA,GCA T31A XP_011542477.1
XM_011544176.2 91 Missense Mutation ACA,GCA T31A XP_011542478.1

View Full Product Details