Product Details

SNP ID
rs41303095
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:231364492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAGGAAGTAGAACAAAACTGA[A/G]TATGGGCGAGGAAAAAACAATAAAC
Phenotype
MIM: 606425 MIM: 616086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGLN1 PubMed Links

Gene Details

Gene
EGLN1
Gene Name
egl-9 family hypoxia inducible factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022051.2 3570 UTR 3 NP_071334.1
XM_005273166.4 3570 UTR 3 XP_005273223.1
XM_005273167.4 3570 UTR 3 XP_005273224.1
Gene
SPRTN
Gene Name
SprT-like N-terminal domain
There are no transcripts associated with this gene.

View Full Product Details