Product Details

SNP ID
rs41284311
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:28203656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCTTCCCCACATATCCCACTCC[C/T]CTCTGACTGCCCCACTCAGTCCTGG
Phenotype
MIM: 173393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC8 PubMed Links

Gene Details

Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014280.2 Intron NP_055095.2
Gene
PTAFR
Gene Name
platelet activating factor receptor
There are no transcripts associated with this gene.

View Full Product Details