Product Details

SNP ID
rs45498698
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:22569327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTCGACCATCCACGGGGACTGG[A/G]GCTGGCTCACGTATCCGGCTCATGG
Phenotype
MIM: 176945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA8 PubMed Links

Gene Details

Gene
EPHA8
Gene Name
EPH receptor A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011540969.1 155 UTR 5 XP_011539271.1
XM_011540970.1 155 UTR 5 XP_011539272.1
XM_011540972.1 155 UTR 5 XP_011539274.1
XM_011540973.1 155 Intron XP_011539275.1
XM_011540975.2 155 Intron XP_011539277.1

View Full Product Details