Product Details
- SNP ID
-
rs41304018
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:248204004 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAGGAAAGGGCAAGTCTGGAGAG[A/T]GAGTTACCTAATAAACTTCATGTTT
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2M3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4916082] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2M3
- Gene Name
- olfactory receptor family 2 subfamily M member 3
There are no transcripts associated with this gene.
View Full Product Details