Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001312871.1 | Intron | NP_001299800.1 | |||
NM_001312872.1 | Intron | NP_001299801.1 | |||
NM_001312873.1 | Intron | NP_001299802.1 | |||
NM_001312874.1 | Intron | NP_001299803.1 | |||
NM_198076.5 | Intron | NP_932342.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004501.3 | Intron | NP_004492.2 | |||
NM_031844.2 | Intron | NP_114032.2 | |||
XM_017001115.1 | Intron | XP_016856604.1 | |||
XM_017001116.1 | Intron | XP_016856605.1 | |||
XM_017001117.1 | Intron | XP_016856606.1 |