Product Details

SNP ID
rs45538634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233636827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCATGCTCAATGGAAAGCACAGG[C/T]ACAAAGTATATTTTCTCTATTAATG
Phenotype
MIM: 606435 MIM: 606433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UGT1A10 PubMed Links

Gene Details

Gene
UGT1A10
Gene Name
UDP glucuronosyltransferase family 1 member A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019075.2 351 Missense Mutation GCA,GTA A102V NP_061948.1
Gene
UGT1A8
Gene Name
UDP glucuronosyltransferase family 1 member A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019076.4 351 Intron NP_061949.3

View Full Product Details