Product Details

SNP ID
rs41265581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:188979578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTAGCTTTAGAGATATAACCCTT[A/C]TTCATAAGGTCAAGAGAAAATAAAC
Phenotype
MIM: 120180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COL3A1 PubMed Links

Gene Details

Gene
COL3A1
Gene Name
collagen type III alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000090.3 Intron NP_000081.1
Gene
MIR1245A
Gene Name
microRNA 1245a
There are no transcripts associated with this gene.

Gene
MIR1245B
Gene Name
microRNA 1245b
There are no transcripts associated with this gene.

View Full Product Details