Product Details

SNP ID
rs41500846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201185421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATTTCTGCCACGTGCTGGAAAT[A/G]AATGTAAGGGCTCCAATCAGGCTAC
Phenotype
MIM: 601762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP10 PubMed Links

Gene Details

Gene
CASP10
Gene Name
caspase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206524.1 Intron NP_001193453.1
NM_001206542.1 Intron NP_001193471.1
NM_001230.4 Intron NP_001221.2
NM_001306083.1 Intron NP_001293012.1
NM_032974.4 Intron NP_116756.2
NM_032976.3 Intron NP_116758.1
NM_032977.3 Intron NP_116759.2
XM_005246907.2 Intron XP_005246964.1
XM_006712796.3 Intron XP_006712859.1
XM_011511990.2 Intron XP_011510292.1

View Full Product Details