Product Details

SNP ID
rs41529948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224471300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCTTAAGTTACAGTAGACAGG[C/G]AAAGATAAAAATCTTTAACACTAGT
Phenotype
MIM: 603136
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CUL3 PubMed Links

Gene Details

Gene
CUL3
Gene Name
cullin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257197.1 5640 UTR 3 NP_001244126.1
NM_001257198.1 5640 UTR 3 NP_001244127.1
NM_003590.4 5640 UTR 3 NP_003581.1
XM_006712800.2 5640 Intron XP_006712863.2
XM_011511994.2 5640 Intron XP_011510296.1
XM_011511995.1 5640 Intron XP_011510297.1
XM_011511996.2 5640 Intron XP_011510298.1

View Full Product Details