Product Details

SNP ID
rs45568040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:47697557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTGATTGGTCCAACCAGAAGC[A/G]CTGATTCTCCCCACCTGATTTCCTG
Phenotype
MIM: 614268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRF2 PubMed Links

Gene Details

Gene
ADGRF2
Gene Name
adhesion G protein-coupled receptor F2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153839.6 4886 UTR 3 NP_722581.4
Gene
ADGRF4
Gene Name
adhesion G protein-coupled receptor F4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153838.3 4886 Intron NP_722580.3

View Full Product Details