Product Details

SNP ID
rs45529733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99446041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGAGCAGAGAATCAGAGCAAA[A/G]GGCTGGTTTTAATGAATGGTTTTGG
Phenotype
MIM: 603052 MIM: 614774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP5J2-PTCD1 PubMed Links

Gene Details

Gene
ATP5J2-PTCD1
Gene Name
ATP5J2-PTCD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198879.1 Intron NP_001185808.1
Gene
CPSF4
Gene Name
cleavage and polyadenylation specific factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081559.2 Intron NP_001075028.1
NM_001318160.1 Intron NP_001305089.1
NM_001318161.1 Intron NP_001305090.1
NM_001318162.1 Intron NP_001305091.1
NM_006693.3 Intron NP_006684.1
XM_011515755.2 Intron XP_011514057.1
XM_011515756.2 Intron XP_011514058.1
XM_011515757.2 Intron XP_011514059.1
XM_017011700.1 Intron XP_016867189.1
XM_017011701.1 Intron XP_016867190.1
XM_017011702.1 Intron XP_016867191.1
XM_017011703.1 Intron XP_016867192.1
Gene
PTCD1
Gene Name
pentatricopeptide repeat domain 1
There are no transcripts associated with this gene.

View Full Product Details