Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004722.3 | Intron | NP_004713.2 | |||
XM_005250689.4 | Intron | XP_005250746.1 | |||
XM_005250690.4 | Intron | XP_005250747.1 | |||
XM_006716175.3 | Intron | XP_006716238.1 | |||
XM_011516685.2 | Intron | XP_011514987.1 | |||
XM_017012790.1 | Intron | XP_016868279.1 | |||
XM_017012791.1 | Intron | XP_016868280.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278595.1 | Intron | NP_001265524.1 | |||
NM_005916.4 | Intron | NP_005907.3 | |||
NM_182776.2 | Intron | NP_877577.1 | |||
XM_005250348.3 | Intron | XP_005250405.1 | |||
XM_017012217.1 | Intron | XP_016867706.1 |