Product Details

SNP ID
rs41322349
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:107250633 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGACAATGTTGCATATTCCAGATT[A/G]ATTAGCTATTAGGTTTTTTGCAGAT
Phenotype
MIM: 601667
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANGPT1 PubMed Links

Gene Details

Gene
ANGPT1
Gene Name
angiopoietin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146.4 2398 UTR 3 NP_001137.2
NM_001199859.2 2398 UTR 3 NP_001186788.1
NM_001314051.1 2398 UTR 3 NP_001300980.1

View Full Product Details