Product Details

SNP ID
rs41274959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:100306684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGGAAGAAATAGCCCAAATGG[A/G]AATTTTAGGTACCTAAGACTTCGGC
Phenotype
MIM: 243305 MIM: 616717
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INVS PubMed Links

Gene Details

Gene
INVS
Gene Name
inversin
There are no transcripts associated with this gene.

Gene
TEX10
Gene Name
testis expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161584.1 Intron NP_001155056.1
NM_017746.3 Intron NP_060216.2
XM_011518798.1 Intron XP_011517100.1
XM_017014848.1 Intron XP_016870337.1

View Full Product Details