Product Details

SNP ID
rs41301517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101361473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACATGCAGTCCTCATATATTCAG[C/T]TGGTTAGCAAACTAAAGTATTTTTT
Phenotype
MIM: 602938
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAAT PubMed Links

Gene Details

Gene
BAAT
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127610.1 2422 UTR 3 NP_001121082.1
NM_001701.3 2422 UTR 3 NP_001692.1

View Full Product Details