Product Details

SNP ID
rs41305499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92838430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGACAGCTAAGTCTGTCTTTTT[C/T]CTTAGGAAATAAAGGTTTTCCTAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD19P PubMed Links

Gene Details

Gene
ANKRD19P
Gene Name
ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF484
Gene Name
zinc finger protein 484
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007101.2 Intron NP_001007102.1
NM_001261458.1 Intron NP_001248387.1
NM_001261459.1 Intron NP_001248388.1
NM_001261460.1 Intron NP_001248389.1
NM_031486.2 Intron NP_113674.1
XM_006717299.3 Intron XP_006717362.1
XM_011519073.2 Intron XP_011517375.1
XM_011519074.2 Intron XP_011517376.1
XM_011519075.2 Intron XP_011517377.1
XM_017015178.1 Intron XP_016870667.1
XM_017015179.1 Intron XP_016870668.1
XM_017015180.1 Intron XP_016870669.1
XM_017015181.1 Intron XP_016870670.1

View Full Product Details