Product Details

SNP ID
rs41316968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127817803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACTGGTGACCATACTACATGGA[C/T]GTGCGGGTGGTTAGATTCCTGGGTG
Phenotype
MIM: 131195 MIM: 136510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENG PubMed Links

Gene Details

Gene
ENG
Gene Name
endoglin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000118.3 Intron NP_000109.1
NM_001114753.2 Intron NP_001108225.1
NM_001278138.1 Intron NP_001265067.1
Gene
FPGS
Gene Name
folylpolyglutamate synthase
There are no transcripts associated with this gene.

Gene
LOC102723566
Gene Name
uncharacterized LOC102723566
There are no transcripts associated with this gene.

View Full Product Details