Product Details

SNP ID
rs41299238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67796974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAAAAGCATTATGAGAACTGA[C/T]TCAAGGATGGAGGAATCAATTAGTA
Phenotype
MIM: 606060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC12 PubMed Links

Gene Details

Gene
DNAJC12
Gene Name
DnaJ heat shock protein family (Hsp40) member C12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021800.2 679 UTR 3 NP_068572.1
NM_201262.1 679 Intron NP_957714.1
XM_017016431.1 679 UTR 3 XP_016871920.1
XM_017016432.1 679 UTR 3 XP_016871921.1

View Full Product Details