Product Details

SNP ID
rs45461901
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30010535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCTATTCCTCCCTCTTCTCCA[A/G]GATGTATCATTTATTTGATATGTAA
Phenotype
MIM: 176266
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCNA4 PubMed Links

Gene Details

Gene
KCNA4
Gene Name
potassium voltage-gated channel subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002233.3 3385 UTR 3 NP_002224.1

View Full Product Details